Please login to your online account to display your discounted pricing

R&D Systems Human His6 UbcH6/UBE2E1 Recombinant Protein

$175.98 - $296.95

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7324
Molecular Weight 23 kDa
Name His6 UbcH6/UBE2E1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2630050

 
r&d systems
RDSE2630050
50μg Each for $175.98

E2630100

 
r&d systems
RDSE2630100
100μg Each for $296.95
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7324
Molecular Weight 23 kDa
Name His6 UbcH6/UBE2E1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal Gly-Ser-Ser and 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 23 kDa
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7324
Regulatory Status: RUO