Please login to your online account to display your discounted pricing

R&D Systems Human His6 UbcH3 Recombinant Protein


Manufacturer: r&d systems  RDSE2610050

 View more versions of this product

Catalog No. E2610050

  • / Each
 

Description & Specifications

Specifications

Source Human
Molecular Weight 28 kDa
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 997
Name His6 UbcH3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Conjugate Unlabeled
Quantity 50μg