Please login to your online account to display your discounted pricing

R&D Systems Human His6 UbcH3 Recombinant Protein


Manufacturer: r&d systems  RDSE2610100

 View more versions of this product

Catalog No. E2610100

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 28 kDa
Name His6 UbcH3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 100μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 997
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.