Please login to your online account to display your discounted pricing

R&D Systems Human His6 UbcH10/UBE2C Recombinant Protein

$172.26 - $286.85

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 11065
Molecular Weight 21 kDa
Name His6 UbcH10/UBE2C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2650050

 
r&d systems
RDSE2650050
50μg Each for $172.26

E2650100

 
r&d systems
RDSE2650100
100μg Each for $286.85
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 11065
Molecular Weight 21 kDa
Name His6 UbcH10/UBE2C
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal Met and 7-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 21 kDa
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 11065
Regulatory Status: RUO