Please login to your online account to display your discounted pricing

R&D Systems Human His6 HR6B/UBE2B Recombinant Protein


Manufacturer: r&d systems  RDSE2613050

 View more versions of this product

Catalog No. E2613050

  • / Each
 

Description & Specifications

Specifications

Name His6 HR6B/UBE2B
Gene ID (Entrez) 7320
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Source Human
Quantity 50μg
Recombinant Yes