missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human His6 HR6B/UBE2B Recombinant Protein

$175.98 - $293.04

Specifications

Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 7320
Name His6 HR6B/UBE2B
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E2613050 R&D Systems
RDSE2613050
50μg Each for $175.98
E2613100 R&D Systems
RDSE2613100
100μg Each for $293.04
Specifications

Description

Source: E. coli-derived, Contains an N-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 7320
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7320
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
No Formulation Info in Bulk Inventory
His6 HR6B/UBE2B
Yes