Please login to your online account to display your discounted pricing

R&D Systems Human His6 HR6A/UBE2A Recombinant Protein


Manufacturer: r&d systems  RDSE2612050

 View more versions of this product

Catalog No. E2612050

  • / Each
 

Description & Specifications

Specifications

Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7319
Name His6 HR6A/UBE2A
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Source Human
Conjugate Unlabeled
Recombinant Yes