Please login to your online account to display your discounted pricing

R&D Systems Human His6 HR6A/UBE2A Recombinant Protein

$186.40 - $298.51

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7319
Name His6 HR6A/UBE2A
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2612100

 
r&d systems
RDSE2612100
100μg Each for $298.51

E2612050

 
r&d systems
RDSE2612050
50μg Each for $186.40
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7319
Name His6 HR6A/UBE2A
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7319
Regulatory Status: RUO