Please login to your online account to display your discounted pricing

R&D Systems Human His6 ATG3 Recombinant Protein


Manufacturer: r&d systems  RDSE2670050

 View more versions of this product

Catalog No. E2670050

  • / Each
 

Description & Specifications

Specifications

Recombinant Yes
Gene ID (Entrez) 64422
Name His6 ATG3
Formulation No Formulation Info in Bulk Inventory
Source Human
Quantity 50μg
Conjugate Unlabeled
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.