missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human His6 ATG3 Recombinant Protein

$175.98 - $296.95

Specifications

Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 64422
Name His6 ATG3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E2670050 R&D Systems
RDSE2670050
50μg Each for $175.98
E2670100 R&D Systems
RDSE2670100
100μg Each for $296.95
Specifications

Description

Source: E. coli-derived, Contains a 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 64422
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
64422
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
No Formulation Info in Bulk Inventory
His6 ATG3
Yes