missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human HGF Recombinant Protein, (NS0-expressed)

$424.39 - $1,387.19

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3082
Molecular Weight alpha chain: 53.7 kDabeta chain: 26 kDa
Name HGF
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
294HGN005 R&D Systems
RDS294HGN005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $424.39
294HGN005CF R&D Systems
RDS294HGN005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $424.39
294HGN025 R&D Systems
RDS294HGN025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $1,387.19
294HGN025CF R&D Systems
RDS294HGN025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $1,387.19
Specifications

Description

  • Source: Mouse myeloma cell line, NS0-derived, Gln32-Ser728
  • Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: alpha chain: 53.7 kDabeta chain: 26 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
  • Entrez Gene ID: 3082
  • Regulatory Status: RUO
Description & Specifications

Specifications

Unlabeled
alpha chain: 53.7 kDabeta chain: 26 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3082
HGF
Yes