Please login to your online account to display your discounted pricing

R&D Systems Human HGF R/c-MET Fc Chimera Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4233
Molecular Weight 32.5 kDa (alpha chain), 96.7 kDa (beta chain)
Name HGF R/c-MET Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

358MT100

 
r&d systems
RDS358MT100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94

358MT100CF

 
r&d systems
RDS358MT100CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $454.94
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4233
Molecular Weight 32.5 kDa (alpha chain), 96.7 kDa (beta chain)
Name HGF R/c-MET Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 100μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Glu25-Thr932 with C-terminal human Fc and a 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 32.5 kDa (alpha chain), 96.7 kDa (beta chain)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 4233
Regulatory Status: RUO