Please login to your online account to display your discounted pricing

R&D Systems Human Heterodimer Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1082
Molecular Weight 15.5 kDa (CG beta) & 10.2 kDa (CG alpha)
Name Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

7727CG010

 
r&d systems
RDS7727CG010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

7727CG010CF

 
r&d systems
RDS7727CG010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1082
Molecular Weight 15.5 kDa (CG beta) & 10.2 kDa (CG alpha)
Name Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser21-Gln165 (CG β) & Ala25-Ser116 (CG α)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa (CG beta) & 10.2 kDa (CG alpha)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1082
Regulatory Status: RUO