Please login to your online account to display your discounted pricing

R&D Systems Human GST UBE2M/Ubc12 Recombinant Protein


Manufacturer: r&d systems  RDSE2655050

 View more versions of this product

Catalog No. E2655050

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9040
Name GST UBE2M/Ubc12
Formulation No Formulation Info in Bulk Inventory
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Source Human
Recombinant Yes
Conjugate Unlabeled
Quantity 50μg