Please login to your online account to display your discounted pricing

R&D Systems Human GST UBE2M/Ubc12 Recombinant Protein

$175.98 - $293.04

Specifications

Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 9040
Name GST UBE2M/Ubc12
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2655050

 
r&d systems
RDSE2655050
50μg Each for $175.98

E2655100

 
r&d systems
RDSE2655100
100μg Each for $293.04
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 9040
Name GST UBE2M/Ubc12
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal GST (glutathione S-transferase) tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 9040
Regulatory Status: RUO