Please login to your online account to display your discounted pricing

R&D Systems Human GST UBE2M/Ubc12 Recombinant Protein


Manufacturer: r&d systems  RDSE2655100

 View more versions of this product

Catalog No. E2655100

  • / Each
 

Description & Specifications

Specifications

Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Source Human
Formulation No Formulation Info in Bulk Inventory
Name GST UBE2M/Ubc12
Gene ID (Entrez) 9040
Recombinant Yes
Conjugate Unlabeled
Quantity 100μg