Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH8 Recombinant Protein


Manufacturer: r&d systems  RDSE2643050

 View more versions of this product

Catalog No. E2643050

  • / Each
 

Description & Specifications

Specifications

Name GST UbcH8
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 9246