Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH5c/UBE2D3 Recombinant Protein


Manufacturer: r&d systems  RDSE2625050

 View more versions of this product

Catalog No. E2625050

  • / Each
 

Description & Specifications

Specifications

Name GST UbcH5c/UBE2D3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 7323