Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH5b/UBE2D2 Recombinant Protein


Manufacturer: r&d systems  RDSE2620050

 View more versions of this product

Catalog No. E2620050

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7322
Name GST UbcH5b/UBE2D2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal GST (glutathione S-transferase) tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7322
Regulatory Status: RUO