Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH5b/UBE2D2 Recombinant Protein


Manufacturer: r&d systems  RDSE2620100

 View more versions of this product

Catalog No. E2620100

  • / Each
 

Description & Specifications

Specifications

Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Name GST UbcH5b/UBE2D2
Recombinant Yes
Source Human
Conjugate Unlabeled
Gene ID (Entrez) 7322
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 100μg