Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH5a/UBE2D1 Recombinant Protein

$172.26 - $286.85

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7321
Molecular Weight 44 kDa
Name GST UbcH5a/UBE2D1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2615050

 
r&d systems
RDSE2615050
50μg Each for $172.26

E2615100

 
r&d systems
RDSE2615100
100μg Each for $286.85
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7321
Molecular Weight 44 kDa
Name GST UbcH5a/UBE2D1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Contains an N-terminal GST (glutathione S-transferase) tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 44 kDa
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7321
Regulatory Status: RUO