Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH5a/UBE2D1 Recombinant Protein


Manufacturer: r&d systems  RDSE2615100

 View more versions of this product

Catalog No. E2615100

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 44 kDa
Name GST UbcH5a/UBE2D1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7321
Quantity 100μg