Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH2/UBE2H Recombinant Protein


Manufacturer: r&d systems  RDSE2605050

 View more versions of this product

Catalog No. E2605050

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7328
Formulation No Formulation Info in Bulk Inventory
Name GST UbcH2/UBE2H
Recombinant Yes
Conjugate Unlabeled
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Source Human
Quantity 50μg