missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human GST UbcH2/UBE2H Recombinant Protein

$171.02 - $284.79

Specifications

Conjugate Unlabeled
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 7328
Name GST UbcH2/UBE2H
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E2605050 R&D Systems
RDSE2605050
50μg Each for $171.02
E2605100 R&D Systems
RDSE2605100
100μg Each for $284.79
Specifications

Description

Source: E. coli-derived, Contains an N-terminal GST (glutathione S-transferase) tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 7328
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7328
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
No Formulation Info in Bulk Inventory
GST UbcH2/UBE2H
Yes