missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Growth Hormone 2 Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2689
Molecular Weight 22.4 kDa
Name Growth Hormone 2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7668GH025 R&D Systems
RDS7668GH025
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $376.73
7668GH025CF R&D Systems
RDS7668GH025CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $376.73
Specifications

Description

Source: E. coli-derived, Phe27-Phe217, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 2689
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2689
Growth Hormone 2
25μg
Human