missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human gp130 Recombinant Protein

$378.79 - $1,240.24

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3572
Molecular Weight 68 kDa
Name gp130
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
228GP010CF R&D Systems
RDS228GP010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $378.79
228GP010 R&D Systems
RDS228GP010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $378.79
228GP050CF R&D Systems
RDS228GP050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $1,240.24
228GP050 R&D Systems
RDS228GP050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $1,240.24
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Leu24-Glu619 (Glu619Asp)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 68 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3572
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
68 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3572
gp130
Yes