missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Glypican 3 Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2719
Molecular Weight 61.6 kDa
Name Glypican 3
Purity 97%, by SDS-PAGE with silver staining.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2119GP050 R&D Systems
RDS2119GP050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $441.90
2119GP050CF R&D Systems
RDS2119GP050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $441.90
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Met1-His559 with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE with silver staining.
Molecular Weight: 61.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2719
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
61.6 kDa
97%, by SDS-PAGE with silver staining.
Yes
2719
Glypican 3
50μg
Human