missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human GFR alpha-3/GDNF R alpha-3 Fc Chimera Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2676
Name GFR alpha-3/GDNF R alpha-3 Fc Chimera
Quantity 100μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Molecular Weight Purity Price Quantity  
Catalog Number Mfr. No. Molecular Weight Purity Price Quantity  
670FR100 R&D Systems
RDS670FR100
67 kDa (monomer) 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $454.94
7269FR100 R&D Systems
RDS7269FR100
64.8 kDa (monomer) 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $454.94
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Asp32-Asn374
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 64.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2676
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
2676
100μg
Human
Lyophilized from a 0.2μm filtered solution in PBS.
GFR alpha-3/GDNF R alpha-3 Fc Chimera
Yes