Please login to your online account to display your discounted pricing

R&D Systems Human Galectin-4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3960
Molecular Weight 36 kDa
Name Galectin-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1227GA050CF

 
r&d systems
RDS1227GA050CF
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl, TCEP, PEG and Trehalose. Each for $428.87

1227GA050

 
r&d systems
RDS1227GA050
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl, TCEP, PEG and Trehalose with BSA as a carrier protein. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3960
Molecular Weight 36 kDa
Name Galectin-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: E. coli-derived, Ala2-Ile323
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in HEPES, NaCl, TCEP, PEG and Trehalose.
Entrez Gene ID: 3960
Regulatory Status: RUO