Please login to your online account to display your discounted pricing

R&D Systems Human Galectin-3 Recombinant Protein


Manufacturer: r&d systems  RDS1154GA050

 View more versions of this product

Catalog No. 1154GA050

  • / Each
 

Description & Specifications

Specifications

Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight 26 kDa
Formulation Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein.
Gene ID (Entrez) 3958
Recombinant Yes
Conjugate Unlabeled
Name Galectin-3
Source Human
Quantity 50μg