missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Galectin-3 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3958
Molecular Weight 26 kDa
Name Galectin-3
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1154GA050 R&D Systems
RDS1154GA050
Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein. Each for $415.83
1154GA050CF R&D Systems
RDS1154GA050CF
Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol. Each for $415.83
Specifications

Description

Source: E. coli-derived, Ala2-Ile250
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Entrez Gene ID: 3958
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
26 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3958
Galectin-3
50μg
Human