missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Galectin-2 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3957
Molecular Weight 14 kDa
Name Galectin-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1153GA050 R&D Systems
RDS1153GA050
Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: E. coli-derived, Met1-Glu132
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Entrez Gene ID: 3957
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
3957
Galectin-2
50μg
Human