missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human G-CSF R/CD114 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1441
Molecular Weight 66.5 kDa
Name G-CSF R/CD114
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
381GR050CF R&D Systems
RDS381GR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
381GR050 R&D Systems
RDS381GR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Glu25-Pro621
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 66.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1441
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
66.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
1441
G-CSF R/CD114
50μg
Human