missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FSH alpha/beta Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2488
Molecular Weight 10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
Name FSH alpha/beta
Purity 95%, by SDS-PAGE with silver staining.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5925FS010 R&D Systems
RDS5925FS010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $441.90
5925FS010CF R&D Systems
RDS5925FS010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $441.90
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ala19-Glu129 & Cys21-Glu129 (alpha); Ala25-Ser116 (beta)
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2488
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
95%, by SDS-PAGE with silver staining.
Yes
2488
FSH alpha/beta
10μg
Human