missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Follistatin 300 aa 30-329 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 10468
Molecular Weight 31 kDa
Name Follistatin 300 aa 30-329
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
669FO025CF R&D Systems
RDS669FO025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $416.79
669FO025 R&D Systems
RDS669FO025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gly30-Asp329, with an N-terminal Met
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 31 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 10468
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
31 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
10468
Follistatin 300 aa 30-329
25μg
Human