missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Flt-3 Ligand Recombinant Protein, (NSO derived)

$194.23 - $624.40

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2323
Molecular Weight 18 kDa
Name Flt-3 Ligand
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
308FKN005CF R&D Systems
RDS308FKN005CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 5μg Each for $194.23
308FKN005 R&D Systems
RDS308FKN005
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 5μg Each for $194.23
308FKN025 R&D Systems
RDS308FKN025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 25μg Each for $624.40
308FKN025CF R&D Systems
RDS308FKN025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 25μg Each for $624.40
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Thr27-Pro185
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 2323
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
2323
Flt-3 Ligand
Yes