missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Flt-3/Flk-2 Fc Chimera Recombinant Protein

$429.46 - $429.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2322
Molecular Weight 85.5 kDa (monomer)
Name Flt-3/Flk-2 Fc Chimera
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
368ST050 R&D Systems
RDS368ST050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $429.46
368ST050CF R&D Systems
RDS368ST050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $429.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Met1-Asn541
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 85.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2322
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
85.5 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2322
Flt-3/Flk-2 Fc Chimera
50μg
Human