missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FGF basic 157 aa Recombinant Protein

For use in cell or tissue culture applications.

$328.11 - $328.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2247
Molecular Weight 17.4 kDa
Name FGF basic 157 aa
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
234FSE025 R&D Systems
RDS234FSE025
Supplied as a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. Each for $328.11
234FSE025CF R&D Systems
RDS234FSE025CF
Supplied as a 0.2μm filtered solution in Tris-HCl and NaCl. Each for $328.11
Specifications

Description

  • Source: E. coli-derived, Gly132-Ser288
  • Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: 17.4 kDa
  • Formulation: Supplied as a 0.2μm filtered solution in Tris-HCl and NaCl.
  • Entrez Gene ID: 2247
  • Regulatory Status: RUO
Description & Specifications

Specifications

Unlabeled
17.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2247
FGF basic 157 aa
25μg
Human