Please login to your online account to display your discounted pricing

R&D Systems Human FGF basic 146 aa Recombinant Protein

$246.37 - $2,613.64

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2247
Molecular Weight 16.5 kDa
Name FGF basic 146 aa
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

233FB025

 
r&d systems
RDS233FB025
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. 25μg Each for $246.37

233FB001MGC

 
r&d systems
RDS233FB001MGCF
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl. 1mg Each for $2,613.64

233FB025CF

 
r&d systems
RDS233FB025CF
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl. 25μg Each for $246.37

23-3FB-01M

 
r&d systems
233FB01M
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein 1mg Each for $2,613.64
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2247
Molecular Weight 16.5 kDa
Name FGF basic 146 aa
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Pro143-Ser288, with an N-terminal Ala
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 2247
Regulatory Status: RUO