missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FGF acidic aa 16-155 Recombinant Protein

$233.33 - $233.33

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2246
Molecular Weight 15.5 kDa
Name FGF acidic aa 16-155
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
232FA025CF R&D Systems
RDS232FA025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA. Each for $233.33
232FA025 R&D Systems
RDS232FA025
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and DTT with BSA as a carrier protein. Each for $233.33
Specifications

Description

Source: E. coli-derived, Phe16-Asp155, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA.
Entrez Gene ID: 2246
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2246
FGF acidic aa 16-155
25μg
Human