Please login to your online account to display your discounted pricing

R&D Systems Human FGF-9 Recombinant Protein

$337.62 - $337.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2254
Molecular Weight 23 kDa
Name FGF-9
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

273F9025CF

 
r&d systems
RDS273F9025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $337.62

273F9025

 
r&d systems
RDS273F9025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $337.62
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2254
Molecular Weight 23 kDa
Name FGF-9
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Met1-Ser208 and Ala2-Ser208
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 23 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2254
Regulatory Status: RUO