missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FGF-8f Recombinant Protein

$328.11 - $328.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2253
Molecular Weight 25.5 kDa
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Name Price Quantity  
Catalog Number Mfr. No. Formulation Name Price Quantity  
5027FF025 R&D Systems
RDS5027FF025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. FGF-8 f Isoform Each for $328.11
5027FF025CF R&D Systems
RDS5027FF025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. FGF-8 Each for $328.11
Specifications

Description

Source: E. coli-derived, Gln23-Arg244, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 2253
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
25.5 kDa
25μg
Human
2253
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes