missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FGF-4 (aa 71-206) Recombinant Protein

$337.62 - $337.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2249
Molecular Weight 15.2 kDa
Name FGF-4 (aa 71-206)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7460F4025 R&D Systems
RDS7460F4025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein. Each for $337.62
7460F4025CF R&D Systems
RDS7460F4025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA. Each for $337.62
Specifications

Description

Source: E. coli-derived, Ser71-Leu206 with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA.
Entrez Gene ID: 2249
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2249
FGF-4 (aa 71-206)
25μg
Human