missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FGF-20 Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 26281
Molecular Weight 24 kDa
Name FGF-20
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2547FG025CF R&D Systems
RDS2547FG025CF
Lyophilized from a 0.2μm filtered solution in MOPS, (NH4)2SO4, DTT and EDTA. Each for $376.73
2547FG025 R&D Systems
RDS2547FG025
Lyophilized from a 0.2μm filtered solution in MOPS, (NH4)2SO4, DTT and EDTA with BSA as a carrier protein. Each for $376.73
Specifications

Description

Source: E. coli-derived, Ala2-Thr211, with an N-terminal Met and a 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, (NH4)2SO4, DTT and EDTA.
Entrez Gene ID: 26281
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
24 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
26281
FGF-20
25μg
Human