missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FGF-16 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8823
Molecular Weight 23.7 kDa
Name FGF-16
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1212FG025 R&D Systems
RDS1212FG025
Lyophilized from a 0.2μm filtered solution in MOPS and Na2SO4 with BSA as a carrier protein. Each for $298.51
1212FG025CF R&D Systems
RDS1212FG025CF
Lyophilized from a 0.2μm filtered solution in MOPS and Na2SO4. Each for $298.51
Specifications

Description

Source: E. coli-derived, Ala2-Arg207
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 23.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS and Na2SO4.
Entrez Gene ID: 8823
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
23.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8823
FGF-16
25μg
Human