missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human FGF-12 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2257
Molecular Weight 20.4 kDa
Name FGF-12
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2246FG025 R&D Systems
RDS2246FG025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA with BSA as a carrier protein. Each for $298.51
2246FG025CF R&D Systems
RDS2246FG025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA. Each for $298.51
Specifications

Description

Source: E. coli-derived, Met1-Thr181
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA.
Entrez Gene ID: 2257
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2257
FGF-12
25μg
Human