missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Fc gamma RIIA/CD32a (R167) Recombinant Protein

$429.46 - $429.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2212
Molecular Weight 22 kDa
Name Fc gamma RIIA/CD32a (R167)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1330CD050 R&D Systems
RDS1330CD050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $429.46
1330CD050CF R&D Systems
RDS1330CD050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $429.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala36-Ile218, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2212
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2212
Fc gamma RIIA/CD32a (R167)
50μg
Human