missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Fas/TNFRSF6/CD95 Fc Chimera Recombinant Protein

$462.76 - $467.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 355
Name Fas/TNFRSF6/CD95 Fc Chimera
Quantity 50μg
Recombinant Yes
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Purity Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Purity Price Quantity  
326FS050 R&D Systems
RDS326FS050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 45 kDa (monomer) 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $462.76
326FS050CF R&D Systems
RDS326FS050CF
Lyophilized from a 0.2μm filtered solution in PBS. 45 kDa (monomer) 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $462.76
7398FS050 R&D Systems
RDS7398FS050
Lyophilized from a 0.2μm filtered solution in PBS. 43.2 kDa (monomer) 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $467.98
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived, Gln26-Asn173
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 43.2 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 355
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
Fas/TNFRSF6/CD95 Fc Chimera
Yes
355
50μg
Human