missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Fas Ligand/TNFSF6 Recombinant Protein

$467.98 - $467.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 356
Molecular Weight 18 kDa
Name Fas Ligand/TNFSF6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
126FL010 R&D Systems
RDS126FL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $467.98
126FL010CF R&D Systems
RDS126FL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $467.98
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Pro134-Leu281, with an N-terminal Met and 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 356
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
356
Fas Ligand/TNFSF6
10μg
Human