Please login to your online account to display your discounted pricing

R&D Systems Human Epigen Recombinant Protein

$292.21 - $292.21

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 255324
Molecular Weight 6 kDa
Name Epigen
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

6629EP025

 
r&d systems
RDS6629EP025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $292.21

6629EP025CF

 
r&d systems
RDS6629EP025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $292.21
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 255324
Molecular Weight 6 kDa
Name Epigen
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Leu54-Ala104, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 255324
Regulatory Status: RUO